α-Cyclodextrin 97%

BD113410-1000G BD113410-100G
BLDPBD113410-1000GEA 298 EUR
BLDPBD113410-1000G BLDPBD113410-100G
α-Cyclodextrin 97%
α-Cyclodextrine

Storage condition: Sealed in dry, Room Temperature

Formula: C₃₆H₆₀O₃₀
MW: 972.84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078207
CAS Number: 10016-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR