1,3-Diphenylisobenzofuran 95%

BD1137-100G BD1137-5G
BLDPBD1137-100GEA 1000 EUR
BLDPBD1137-100G BLDPBD1137-5G
1,3-Diphenylisobenzofuran 95%
1,3-Diphenylisobenzofuran

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₂₀H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00005931
CAS Number: 5471-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR