Amyl-2-methylbutyrate 98%

BD114275-100G BD114275-500G
BLDPBD114275-100GEA 94.5 EUR
BLDPBD114275-100G BLDPBD114275-500G
Amyl-2-methylbutyrate 98%
Amyl-2-methylbutyrate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00129713
CAS Number: 68039-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR