2,3-Diphenyl-1H-indole 97%

BD1144-5G BD1144-25G
BLDPBD1144-5GEA 133 EUR
BLDPBD1144-5G BLDPBD1144-25G
2,3-Diphenyl-1H-indole 97%
2,3-Diphenyl-1H-indole

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₀H₁₅N
Storage Temperature: Ambient
MDL Number: MFCD00022698
CAS Number: 3469-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR