Benzo[b]thiophene-2-carboxaldehyde 98%

BD11541-1G BD11541-10G
BLDPBD11541-1GEA 9.5 EUR
BLDPBD11541-1G BLDPBD11541-10G
Benzo[b]thiophene-2-carboxaldehyde 98%
Benzo[b]thiophene-2-carboxaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₉H₆OS
MW: 162.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01075041
CAS Number: 3541-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR