Benzanilide 98%

BD115417-1000G BD115417-25G
BLDPBD115417-1000GEA 292 EUR
BLDPBD115417-1000G BLDPBD115417-25G
Benzanilide 98%
Benzanilide

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Boiling Pt: 247
Storage Temperature: Ambient
MDL Number: MFCD00003069
CAS Number: 93-98-1
Merck Index: 13,01060

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR