Benzofuran-7-carboxylic acid 95%

BD115763-5G BD115763-25G BD115763-100MG
BLDPBD115763-5GEA 730 EUR
BLDPBD115763-5G BLDPBD115763-25G BLDPBD115763-100MG
Benzofuran-7-carboxylic acid 95%
Benzofuran-7-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₆O₃
MW: 162.142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10000617
CAS Number: 90484-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR