3-Bromo-2-fluorobenzonitrile 98%

BD115860-250MG BD115860-1G
BLDPBD115860-250MGEA 6.3 EUR
BLDPBD115860-250MG BLDPBD115860-1G
3-Bromo-2-fluorobenzonitrile 98%
3-Bromo-2-fluorobenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₃BrFN
Storage Temperature: Ambient
MDL Number: MFCD06657980
CAS Number: 840481-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR