N,N-Bis(isopropyl)benzylamine 98%

BD116276-100G BD116276-25G
BLDPBD116276-100GEA 779 EUR
BLDPBD116276-100G BLDPBD116276-25G
N,N-Bis(isopropyl)benzylamine 98%
N,N-Bis(isopropyl)benzylamine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₃H₂₁N
Storage Temperature: Ambient
MDL Number: MFCD07368226
CAS Number: 34636-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR