(R)-4-Phenyloxazolidine-2-thione 98+%
|
|
BD11727-100MG
BD11727-5G
BLDPBD11727-100MGEA
20
EUR
BLDPBD11727-100MG
BLDPBD11727-5G
(R)-4-Phenyloxazolidine-2-thione 98+%
(R)-4-Phenyloxazolidine-2-thione
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₉H₉NOS Storage Temperature: Ambient |
MDL Number:
MFCD06658220 CAS Number: 171877-37-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...