(R)-3-(Boc-amino)-3-(2-bromophenyl)propionic acid 98%
|
|
BD117505-250MG
BD117505-100MG
BLDPBD117505-250MGEA
65.1
EUR
BLDPBD117505-250MG
BLDPBD117505-100MG
(R)-3-(Boc-amino)-3-(2-bromophenyl)propionic acid 98%
(R)-3-(Boc-amino)-3-(2-bromophenyl)propionic acid
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₄H₁₈BrNO₄ Storage Temperature: Ambient |
MDL Number:
MFCD03427961 CAS Number: 500789-07-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...