Fmoc-3-(Boc-aminomethyl)-L-phenylalanine 95%
|
|
BD118169-250MG
BD118169-100MG
BLDPBD118169-250MGEA
175
EUR
BLDPBD118169-250MG
BLDPBD118169-100MG
Fmoc-3-(Boc-aminomethyl)-L-phenylalanine 95%
Fmoc-3-(Boc-aminomethyl)-L-phenylalanine
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₃₀H₃₂N₂O₆ MW: 516.58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01860734 CAS Number: 266999-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...