2-Allyl-3-hydroxybenzaldehyde 97% stabilised


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

BD11820-1G BD11820-100MG
BLDPBD11820-1GEA 195 EUR
BLDPBD11820-1G BLDPBD11820-100MG
2-Allyl-3-hydroxybenzaldehyde 97% stabilised
2-Allyl-3-hydroxybenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₁₀O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00100646
CAS Number: 79950-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR