N-(tert-Butoxycarbonyl)-L-methioninol 97%
|
|
BD118326-1G
BD118326-5G
BLDPBD118326-1GEA
16.8
EUR
BLDPBD118326-1G
BLDPBD118326-5G
N-(tert-Butoxycarbonyl)-L-methioninol 97%
N-(tert-Butoxycarbonyl)-L-methioninol
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₁₀H₂₁NO₃S MW: 235.34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00235939 CAS Number: 51372-93-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...