N-Boc-trans-4-fluoro-L-proline methyl ester 97%

BD118476-5G BD118476-25G BD118476-10G
BLDPBD118476-5GEA 88.2 EUR
BLDPBD118476-5G BLDPBD118476-25G BLDPBD118476-10G
N-Boc-trans-4-fluoro-L-proline methyl ester 97%
N-Boc-trans-4-fluoro-L-proline methyl ester

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₁H₁₈FNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD07367993
CAS Number: 203866-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR