4-Methoxyphenylacetone 98%

BD12017-500G BD12017-100G
BLDPBD12017-500GEA 182 EUR
BLDPBD12017-500G BLDPBD12017-100G
4-Methoxyphenylacetone 98%
4-Methoxyphenylacetone

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 268
Melting Pt: -15
Storage Temperature: Ambient
MDL Number: MFCD00008773
CAS Number: 122-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR