1-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid 95%

BD120185-1G BD120185-250MG
BLDPBD120185-1GEA 141 EUR
BLDPBD120185-1G BLDPBD120185-250MG
1-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid 95%
1-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₃H₁₅NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD03426163
CAS Number: 190004-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR