D-Alanine benzyl ester-p-toluenesulphonate 95%

BD12162-500G BD12162-10G
BLDPBD12162-500GEA 777 EUR
BLDPBD12162-500G BLDPBD12162-10G
D-Alanine benzyl ester-p-toluenesulphonate 95%
D-Alanine benzyl ester-p-toluenesulphonate

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₀H₁₃NO₂
Storage Temperature: Freezer
MDL Number: MFCD00039092
CAS Number: 41036-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR