Boc-L-cyclopropylglycine 97%

BD12356-5G BD12356-100MG
BLDPBD12356-5GEA 218 EUR
BLDPBD12356-5G BLDPBD12356-100MG
Boc-L-cyclopropylglycine 97%
Boc-L-cyclopropylglycine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₀H₁₇NO₄
MW: 215.249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659132
CAS Number: 155976-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR