Di(2-pyridyl) ketone 97%
|
|
BD124013-100G
BD124013-5G
BLDPBD124013-100GEA
478
EUR
BLDPBD124013-100G
BLDPBD124013-5G
Di(2-pyridyl) ketone 97%
Di(2-pyridyl) ketone
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₁H₈N₂O Storage Temperature: Ambient |
MDL Number:
MFCD00006288 CAS Number: 19437-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...