Dimethyl 2,5-dibromoterephthalate 97%

BD125305-1G BD125305-100G
BLDPBD125305-1GEA 25.2 EUR
BLDPBD125305-1G BLDPBD125305-100G
Dimethyl 2,5-dibromoterephthalate 97%
Dimethyl 2,5-dibromoterephthalate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₈Br₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00204592
CAS Number: 18014-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR