Dimethyl ethylmalonate 95%
|
|
BD125406-100G
BD125406-25G
BLDPBD125406-100GEA
42
EUR
BLDPBD125406-100G
BLDPBD125406-25G
Dimethyl ethylmalonate 95%
Dimethyl ethylmalonate
Storage condition: Sealed in dry, Room Temperature
Formula:
C₇H₁₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00042839 CAS Number: 26717-67-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...