4-Quinolinecarbaldehyde 98%

BD12614-10G BD12614-25G
BLDPBD12614-10GEA 84 EUR
BLDPBD12614-10G BLDPBD12614-25G
4-Quinolinecarbaldehyde 98%
4-Quinolinecarbaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₀H₇NO
Storage Temperature: Ambient
MDL Number: MFCD00006781
CAS Number: 4363-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR