Ethyl-1-methyl-1H-imidazole-2-carboxylate 98%

BD127898-10G BD127898-25G
BLDPBD127898-10GEA 50.4 EUR
BLDPBD127898-10G BLDPBD127898-25G
Ethyl-1-methyl-1H-imidazole-2-carboxylate 98%
Ethyl-1-methyl-1H-imidazole-2-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₁₀N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD02683326
CAS Number: 30148-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR