3-(Ethoxycarbonyl)phenylboronic acid pinacol ester 98%
|
|
BD128457-1G
BD128457-10G
BLDPBD128457-1GEA
13.7
EUR
BLDPBD128457-1G
BLDPBD128457-10G
3-(Ethoxycarbonyl)phenylboronic acid pinacol ester 98%
3-(Ethoxycarbonyl)phenylboronic acid pinacol ester
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₅H₂₁BO₄ Storage Temperature: Ambient |
MDL Number:
MFCD03093896 CAS Number: 269410-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...