Ethyl-2-(1H-indol-2-yl)acetate 97%

BD129367-250MG BD129367-1G
BLDPBD129367-250MGEA 17.9 EUR
BLDPBD129367-250MG BLDPBD129367-1G
Ethyl-2-(1H-indol-2-yl)acetate 97%
Ethyl-2-(1H-indol-2-yl)acetate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₂H₁₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08361732
CAS Number: 33588-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR