Ethyl-3-iodopropanoate 96%

BD129683-1G BD129683-10G
BLDPBD129683-1GEA 26.3 EUR
BLDPBD129683-1G BLDPBD129683-10G
Ethyl-3-iodopropanoate 96%
Ethyl-3-iodopropanoate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₅H₉IO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00045323
CAS Number: 6414-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR