N-(Piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide 95%

BD130409-1G BD130409-250MG
BLDPBD130409-1GEA 51.5 EUR
BLDPBD130409-1G BLDPBD130409-250MG
N-(Piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide 95%
N-(Piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₇H₂₀F₆N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00864713
CAS Number: 54143-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR