Fmoc-D-Cys(Acm)-OH 97%
|
|
BD131065-1G
BD131065-25G
BLDPBD131065-1GEA
15.8
EUR
BLDPBD131065-1G
BLDPBD131065-25G
Fmoc-D-Cys(Acm)-OH 97%
Fmoc-D-Cys(Acm)-OH
Storage condition: Sealed in dry, 2-8°C
Formula:
C₂₁H₂₂N₂O₅S Storage Temperature: Refrigerator |
MDL Number:
MFCD00077053 CAS Number: 168300-88-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...