H-D-Asp-OMe 95%
|
|
BD132836-10G
BD132836-25G
BLDPBD132836-10GEA
80.9
EUR
BLDPBD132836-10G
BLDPBD132836-25G
H-D-Asp-OMe 95%
H-D-Asp-OMe
Storage condition: Sealed in dry, Room Temperature
Formula:
C₅H₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD03424227 CAS Number: 65414-78-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...