Hexyl chloroformate 95%

BD133349-5G BD133349-1G
BLDPBD133349-5GEA 11.6 EUR
BLDPBD133349-5G BLDPBD133349-1G
Hexyl chloroformate 95%
Hexyl chloroformate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₁₃ClO₂
MW: 164.6299 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000651
CAS Number: 6092-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR