H-Tyr(tBu)-OMe·HCl 95%

BD133973-5G BD133973-100G
BLDPBD133973-5GEA 36.8 EUR
BLDPBD133973-5G BLDPBD133973-100G
H-Tyr(tBu)-OMe·HCl 95%
H-Tyr(tBu)-OMe·HCl

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₄H₂₂ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00153466
CAS Number: 51482-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR