1-Boc-3-cyanopiperidine 97%
|
|
BD13436-25G
BD13436-10G
BLDPBD13436-25GEA
101
EUR
BLDPBD13436-25G
BLDPBD13436-10G
1-Boc-3-cyanopiperidine 97%
1-Boc-3-cyanopiperidine
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₁H₁₈N₂O₂ MW: 210.27 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01861222 CAS Number: 91419-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...