Indium(III) iodide 99.999%
|
|
BD134587-5G
BD134587-25G
BLDPBD134587-5GEA
116
EUR
BLDPBD134587-5G
BLDPBD134587-25G
Indium(III) iodide 99.999%
Indium (III) iodide
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
I₃In MW: 495.53 g/mol Boiling Pt: 500 Melting Pt: 209.85 Storage Temperature: Ambient |
MDL Number:
MFCD00016151 CAS Number: 13510-35-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...