N-(ɑ)-Boc-L-lysine 98%

BD13518-500G BD13518-5G
BLDPBD13518-500GEA 806 EUR
BLDPBD13518-500G BLDPBD13518-5G
N-(ɑ)-Boc-L-lysine 98%
N-(alpha)-Boc-L-lysine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₁H₂₂N₂O₄
MW: 246.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038203
CAS Number: 13734-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR