L-Cystine disodium salt 99%

BD136051-1000G BD136051-10G
BLDPBD136051-1000GEA 282 EUR
BLDPBD136051-1000G BLDPBD136051-10G
L-Cystine disodium salt 99%
L-Cystine disodium salt

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₁₀N₂Na₂O₄S₂
Storage Temperature: Ambient
MDL Number: MFCD00037206
CAS Number: 64704-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR