Calcium-L-threonate 95%

BD136793-1000G BD136793-100G
BLDPBD136793-1000GEA 137 EUR
BLDPBD136793-1000G BLDPBD136793-100G
Calcium-L-threonate 95%
Calcium-L-threonate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₄H₈O₅
Storage Temperature: Ambient
MDL Number: MFCD00077882
CAS Number: 70753-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR