Methyl-2-(benzylamino)acetate 98%
|
|
BD138020-250MG
BD138020-25G
BLDPBD138020-250MGEA
8.4
EUR
BLDPBD138020-250MG
BLDPBD138020-25G
Methyl-2-(benzylamino)acetate 98%
Methyl-2-(benzylamino)acetate
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₁₀H₁₃NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD03247461 CAS Number: 53386-64-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...