Methyl 3-(trimethylsilyl)-4-pentenoate 97%
|
|
BD138469-1G
BD138469-10G
BLDPBD138469-1GEA
20
EUR
BLDPBD138469-1G
BLDPBD138469-10G
Methyl 3-(trimethylsilyl)-4-pentenoate 97%
Methyl 3-(trimethylsilyl)-4-pentenoate
Storage condition: Sealed in dry, 2-8°C
Formula:
C₉H₁₈O₂Si Storage Temperature: Refrigerator |
MDL Number:
MFCD01863734 CAS Number: 185411-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...