Methyl-3-amino-5,6-dichloropyrazine-2-carboxylate 97%
|
|
BD138588-250MG
BD138588-100G
BLDPBD138588-250MGEA
10.5
EUR
BLDPBD138588-250MG
BLDPBD138588-100G
Methyl-3-amino-5,6-dichloropyrazine-2-carboxylate 97%
Methyl 3-amino-5,6-dichloropyrazine-2-carboxylate
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₆H₅Cl₂N₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00010431 CAS Number: 1458-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...