Boc-Bpa-OH 98%

BD1392-1G BD1392-250MG
BLDPBD1392-1GEA 75.6 EUR
BLDPBD1392-1G BLDPBD1392-250MG
Boc-Bpa-OH 98%
Boc-Bpa-OH

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₁H₂₃NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00151886
CAS Number: 104504-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR