Ethyl-5-methylindole-2-carboxylate 97%

BD13936-1G BD13936-10G
BLDPBD13936-1GEA 18.9 EUR
BLDPBD13936-1G BLDPBD13936-10G
Ethyl-5-methylindole-2-carboxylate 97%
Ethyl-5-methylindole-2-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00022703
CAS Number: 16382-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR