6,7-Dimethoxy-1-tetralone 97%
|
|
BD14011-1G
BD14011-100MG
BLDPBD14011-1GEA
92.4
EUR
BLDPBD14011-1G
BLDPBD14011-100MG
6,7-Dimethoxy-1-tetralone 97%
6,7-Dimethoxy-1-tetralone
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₂H₁₄O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00134100 CAS Number: 13575-75-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...