N-(4-Hydroxyphenyl)maleimide 98%
|
|
BD141406-5G
BD141406-10G
BLDPBD141406-5GEA
20
EUR
BLDPBD141406-5G
BLDPBD141406-10G
N-(4-Hydroxyphenyl)maleimide 98%
N-(4-Hydroxyphenyl)maleimide
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₀H₇NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00129167 CAS Number: 7300-91-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...