N-Butylphthalimide 98+%

BD141644-25G BD141644-5G
BLDPBD141644-25GEA 16.8 EUR
BLDPBD141644-25G BLDPBD141644-5G
N-Butylphthalimide 98+%
N-Butylphthalimide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₂H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00039695
CAS Number: 1515-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR