Boc-D-Thi-OH 95%
|
|
BD14205-1G
BD14205-5G
BLDPBD14205-1GEA
38.9
EUR
BLDPBD14205-1G
BLDPBD14205-5G
Boc-D-Thi-OH 95%
Boc-D-Thi-OH
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formula:
C₁₂H₁₇NO₄S Storage Temperature: Refrigerator |
MDL Number:
MFCD00065591 CAS Number: 78452-55-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...