2-Aminophenylboronic acid pinacol ester 98%

BD14210-250MG BD14210-25G
BLDPBD14210-250MGEA 8.4 EUR
BLDPBD14210-250MG BLDPBD14210-25G
2-Aminophenylboronic acid pinacol ester 98%
2-Aminophenylboronic acid pinacol ester

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₂H₁₈BNO₂
Storage Temperature: Ambient
MDL Number: MFCD02179448
CAS Number: 191171-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR