1,4-Bis(2-hydroxyisopropyl)benzene 98%
|
|
BD14233-25G
BD14233-1G
BLDPBD14233-25GEA
215
EUR
BLDPBD14233-25G
BLDPBD14233-1G
1,4-Bis(2-hydroxyisopropyl)benzene 98%
1,4-Bis(2-hydroxyisopropyl)benzene
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₂H₁₈O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00009827 CAS Number: 2948-46-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...