Acenaphthenequinone 98%

BD14338-500G BD14338-100G
BLDPBD14338-500GEA 1470 EUR
BLDPBD14338-500G BLDPBD14338-100G
Acenaphthenequinone 98%
Acenaphthenequinone

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00003805
CAS Number: 82-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR