N-Boc-1,3-propanediamine hydrochloride 98%
|
|
BD143579-250MG
BD143579-1G
BLDPBD143579-250MGEA
11.6
EUR
BLDPBD143579-250MG
BLDPBD143579-1G
N-Boc-1,3-propanediamine hydrochloride 98%
N-Boc-1,3-propanediamine hydrochloride
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₈H₁₉ClN₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD01076598 CAS Number: 127346-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...